Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278083.1 | NP_001265012.1 | 3 - 4 | 352 | 73 | |
GenBank mRNA | AJ851408.1 | - | 1 - 2 | 504 | 73 | |
AM072079.1 | - | 3 - 4 | 364 | 73 | ||
CR407233.1 | - | 3 - 4 | 225 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |