Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012872.2 | NP_001012890.1 | 5 - 6 | 736 | 122 | |
XM_015278047.2 | XP_015133533.1 | 6 - 7 | 122 | |||
XM_015278048.2 | XP_015133534.1 | 6 - 7 | 122 | |||
XM_015278049.2 | XP_015133535.1 | 5 - 6 | 122 | |||
XM_015278050.2 | XP_015133536.1 | 5 - 6 | 122 | |||
XM_015278051.2 | XP_015133537.1 | 5 - 6 | 122 | |||
XM_025148641.1 | XP_025004409.1 | 5 - 6 | 122 | |||
XM_025148642.1 | XP_025004410.1 | 5 - 6 | 122 | |||
GenBank mRNA | AJ719997.1 | - | 5 - 6 | 737 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |