Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012906.1 | NP_001012924.1 | 4 - 5 | 800 | 87 | |
GenBank mRNA | AJ720593.1 | - | 4 - 5 | 800 | 87 | |
CR391018.1 | - | 4 - 5 | 617 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |