Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030866.1 | NP_001026037.1 | 3 - 4 | 394 | 111 | |
GenBank mRNA | AJ720406.1 | - | 3 - 4 | 391 | 111 | |
AJ851573.1 | - | 3 - 4 | 394 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |