Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031077.1 | NP_001026248.1 | 4 - 5 | 724 | 75 | |
GenBank mRNA | AJ719270.1 | - | 4 - 5 | 724 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |