Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031207.2 | NP_001026378.2 | 5 - 6 | 734 | 76 | |
GenBank mRNA | AB264607.1 | - | 5 - 6 | 530 | 76 | |
AJ851709.1 | - | 5 - 6 | 734 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |