Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031221.1 | NP_001026392.1 | 8 - 9 | 1303 | 74 | |
GenBank mRNA | AJ851457.1 | - | 8 - 9 | 1303 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |