Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001195.2 | NP_001001195.1 | 4 - 5 | 1040 | 57 | |
GenBank mRNA | AB734684.1 | - | 4 - 5 | 948 | 57 | |
AB734685.1 | - | 4 - 5 | 948 | 57 | ||
AB734686.1 | - | 4 - 5 | 948 | 57 | ||
AB734687.1 | - | 4 - 5 | 948 | 57 | ||
AY574983.1 | - | 4 - 5 | 1040 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |