Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001203.1 | NP_001001203.1 | 2 - 3 | 232 | 67 | |
GenBank mRNA | AY582132.1 | - | 2 - 3 | 232 | 67 | |
BX930141.1 | - | 2 - 3 | 190 | 67 | ||
BX935342.2 | - | 2 - 3 | 234 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |