Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001529.1 | NP_001001529.1 | 9 - 10 | 1445 | 127 | |
GenBank mRNA | AY463528.1 | - | 9 - 10 | 1445 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |