Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001530.1 | NP_001001530.1 | 6 - 7 | 1145 | 101 | |
XM_015291384.2 | XP_015146870.1 | 6 - 7 | 1229 | 101 | ||
XM_015291385.2 | XP_015146871.1 | 6 - 7 | 836 | 101 | ||
GenBank mRNA | AY463529.1 | - | 6 - 7 | 1145 | 101 | |
CR733175.1 | - | 4 - 5 | 391 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |