Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006464.2 | NP_001006464.1 | 3 - 4 | 330 | 94 | |
GenBank mRNA | AJ720522.1 | - | 3 - 4 | 330 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |