Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204743.2 | NP_990074.2 | 1 - 2 | 437 | 97 | |
XM_025143948.1 | XP_024999716.1 | 2 - 3 | 3393 | 97 | ||
GenBank mRNA | X52749.1 | - | 53 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |