Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204857.1 | NP_990188.1 | 5 - 6 | 657 | 90 | |
XM_015299252.2 | XP_015154738.1 | 5 - 6 | 735 | 90 | ||
GenBank mRNA | AF082666.1 | - | 5 - 6 | 657 | 90 | |
AM049249.1 | - | 5 - 6 | 852 | 90 | ||
AM049250.1 | - | 6 - 7 | 925 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |