Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204944.2 | NP_990275.1 | 4 - 5 | 1370 | 61 | |
XM_015294136.2 | XP_015149622.1 | 4 - 5 | 1600 | 61 | ||
XM_015294137.2 | XP_015149623.1 | 4 - 5 | 1603 | 61 | ||
XM_025155049.1 | XP_025010817.1 | 5 - 6 | 1458 | 61 | ||
XM_025155050.1 | XP_025010818.1 | 5 - 6 | 1455 | 61 | ||
GenBank mRNA | AF009012.1 | - | 4 - 5 | 1370 | 61 | |
AY687628.1 | - | 3 - 4 | 1113 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |