Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4Y1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205108.1 | NP_990439.1 | 4 - 5 | 539 | 59 | |
GenBank mRNA | BX930717.1 | - | 4 - 5 | 492 | 59 | |
BX934312.1 | - | 4 - 5 | 451 | 59 | ||
CR391662.1 | - | 4 - 5 | 543 | 59 | ||
L24368.1 | - | 4 - 5 | 539 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |