Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190165.4 | NP_001177094.1 | 2 - 3 | 200 | 104 | |
NM_205260.5 | NP_990591.1 | 2 - 3 | 200 | 104 | ||
NR_073182.3 | 2 - 3 | 189 | 104 | |||
XM_015289639.2 | XP_015145125.1 | 3 - 4 | 3971 | 104 | ||
GenBank mRNA | DQ185930.1 | - | 2 - 3 | 200 | 104 | |
DQ185931.1 | - | 2 - 3 | 200 | 104 | ||
DR429295.1 | - | 2 - 3 | 196 | 104 | ||
S78477.1 | - | 2 - 3 | 198 | 104 | ||
Y07539.1 | - | 190 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |