Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030549.1 | NP_001025720.1 | 3 - 4 | 1506 | 100 | |
XM_015291905.2 | XP_015147391.1 | 3 - 4 | 1512 | 100 | ||
XM_015291906.2 | XP_015147392.1 | 3 - 4 | 1564 | 100 | ||
XM_015291907.2 | XP_015147393.1 | 3 - 4 | 1493 | 100 | ||
XM_015291908.2 | XP_015147394.1 | 4 - 5 | 1516 | 100 | ||
XM_015291909.2 | XP_015147395.1 | 3 - 4 | 1601 | 100 | ||
XM_015291910.2 | XP_015147396.1 | 3 - 4 | 1378 | 100 | ||
XM_015291911.2 | XP_015147397.1 | 3 - 4 | 1700 | 100 | ||
XM_015291913.2 | XP_015147399.1 | 3 - 4 | 1376 | 100 | ||
XM_015291914.2 | XP_015147400.1 | 3 - 4 | 1512 | 100 | ||
XM_025153831.1 | XP_025009599.1 | 3 - 4 | 1916 | 100 | ||
XM_025153832.1 | XP_025009600.1 | 4 - 5 | 1550 | 100 | ||
GenBank mRNA | AJ851537.1 | - | 3 - 4 | 1506 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |