Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030570.1 | NP_001025741.1 | 6 - 7 | 2555 | 87 | |
GenBank mRNA | AJ719785.1 | - | 6 - 7 | 2555 | 87 | |
CR386283.1 | - | 5 - 6 | 888 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |