Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030597.2 | NP_001025768.1 | 14 - 15 | 1586 | 71 | |
GenBank mRNA | AJ851498.1 | - | 14 - 15 | 1587 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |