Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030597.2 | NP_001025768.1 | 18 - 19 | 1954 | 65 | |
GenBank mRNA | AJ851498.1 | - | 18 - 19 | 1955 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |