Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030616.1 | NP_001025787.1 | 5 - 6 | 721 | 131 | |
XM_015293767.1 | XP_015149253.1 | 5 - 6 | 672 | 131 | ||
XM_015293768.1 | XP_015149254.1 | 4 - 5 | 337 | 131 | ||
GenBank mRNA | AJ720720.1 | - | 5 - 6 | 721 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |