Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030793.3 | NP_001025964.2 | 2 - 3 | 405 | 152 | |
GenBank mRNA | AJ851403.1 | - | 2 - 3 | 405 | 152 | |
BM491256.1 | - | 2 - 3 | 375 | 152 | ||
BU440235.1 | - | 2 - 3 | 385 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |