Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030916.1 | NP_001026087.1 | 690 | 78 | ||
GenBank mRNA | AJ719799.1 | - | 690 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus |