Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030923.1 | NP_001026094.1 | 830 | 95 | ||
XM_015298954.2 | XP_015154440.1 | 793 | 95 | |||
XM_015298955.2 | XP_015154441.1 | 1189 | 95 | |||
XM_025143708.1 | XP_024999476.1 | 980 | 95 | |||
GenBank mRNA | AJ719313.1 | - | 830 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus |