Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031114.1 | NP_001026285.1 | 3 - 4 | 371 | 101 | |
XM_025149546.1 | XP_025005314.1 | 3 - 4 | 409 | 101 | ||
GenBank mRNA | AJ720103.1 | - | 3 - 4 | 371 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |