Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031138.2 | NP_001026309.1 | 3 - 4 | 363 | 89 | |
XM_015276241.2 | XP_015131727.1 | 3 - 4 | 485 | 89 | ||
XM_025149636.1 | XP_025005404.1 | 3 - 4 | 485 | 89 | ||
GenBank mRNA | AY761140.1 | - | 3 - 4 | 364 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |