Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031138.2 | NP_001026309.1 | 4 - 5 | 576 | 96 | |
XM_015276241.2 | XP_015131727.1 | 4 - 5 | 698 | 96 | ||
GenBank mRNA | AY761140.1 | - | 4 - 5 | 577 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |