Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031465.1 | NP_001026636.1 | 2 - 3 | 126 | 60 | |
GenBank mRNA | AJ720828.1 | - | 2 - 3 | 126 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |