Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031531.1 | NP_001026702.1 | 4 - 5 | 937 | 153 | |
GenBank mRNA | AJ720204.1 | - | 4 - 5 | 937 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |