Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030647.1 | NP_001025818.1 | 2 - 3 | 350 | 92 | |
GenBank mRNA | AJ719747.1 | - | 2 - 3 | 350 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |