Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030647.1 | NP_001025818.1 | 5 - 6 | 514 | 139 | |
GenBank mRNA | AJ719747.1 | - | 5 - 6 | 514 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |