Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030654.2 | NP_001025825.2 | 7 - 8 | 659 | 86 | |
XM_015275241.2 | XP_015130727.1 | 6 - 7 | 847 | 86 | ||
XM_025155434.1 | XP_025011202.1 | 5 - 6 | 880 | 86 | ||
XM_025155435.1 | XP_025011203.1 | 6 - 7 | 3849 | 86 | ||
XM_025155436.1 | XP_025011204.1 | 7 - 8 | 4418 | 86 | ||
XM_025155437.1 | XP_025011205.1 | 6 - 7 | 797 | 86 | ||
GenBank mRNA | AJ851383.1 | - | 7 - 8 | 659 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |