Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ721085.1 | - | 2 - 3 | 170 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |