Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030763.2 | NP_001025934.2 | 2 - 3 | 248 | 65 | |
XM_025148736.1 | XP_025004504.1 | 2 - 3 | 241 | 65 | ||
XM_025148737.1 | XP_025004505.1 | 2 - 3 | 251 | 65 | ||
GenBank mRNA | AJ720065.1 | - | 2 - 3 | 248 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |