Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030766.1 | NP_001025937.1 | 2 - 3 | 377 | 95 | |
XM_015285715.2 | XP_015141201.1 | 2 - 3 | 429 | 95 | ||
XM_015285722.2 | XP_015141208.1 | 2 - 3 | 190 | 95 | ||
XM_025148812.1 | XP_025004580.1 | 2 - 3 | 414 | 95 | ||
GenBank mRNA | AJ720327.1 | - | 2 - 3 | 377 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |