Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030789.1 | NP_001025960.1 | 2 - 3 | 258 | 65 | |
XM_015296322.2 | XP_015151808.2 | 3 - 4 | 360 | 65 | ||
XM_015296326.2 | XP_015151812.2 | 2 - 3 | 128 | 65 | ||
XM_025149389.1 | XP_025005157.1 | 3 - 4 | 361 | 65 | ||
GenBank mRNA | AJ719861.1 | - | 2 - 3 | 258 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |