Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030789.1 | NP_001025960.1 | 5 - 6 | 625 | 67 | |
XM_015296322.2 | XP_015151808.2 | 6 - 7 | 727 | 67 | ||
XM_015296326.2 | XP_015151812.2 | 5 - 6 | 495 | 67 | ||
XM_015296328.2 | XP_015151814.2 | 4 - 5 | 583 | 67 | ||
XM_025149388.1 | XP_025005156.1 | 5 - 6 | 638 | 67 | ||
GenBank mRNA | AJ719861.1 | - | 5 - 6 | 625 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |