Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031160.1 | NP_001026331.1 | 6 - 7 | 782 | 89 | |
XM_015285719.2 | XP_015141205.1 | 6 - 7 | 767 | 89 | ||
XM_015285720.2 | XP_015141206.1 | 6 - 7 | 876 | 89 | ||
GenBank mRNA | AJ720429.1 | - | 6 - 7 | 782 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |