Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031277.1 | NP_001026448.1 | 3 - 4 | 566 | 131 | |
GenBank mRNA | AJ851595.1 | - | 3 - 4 | 566 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |