Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031281.1 | NP_001026452.1 | 2 - 3 | 287 | 96 | |
GenBank mRNA | AJ719913.1 | - | 2 - 3 | 287 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |