Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031293.1 | NP_001026464.1 | 5 - 6 | 929 | 103 | |
XM_015291044.2 | XP_015146530.1 | 5 - 6 | 968 | 103 | ||
XM_015291045.2 | XP_015146531.1 | 5 - 6 | 944 | 103 | ||
XM_015291046.2 | XP_015146532.1 | 5 - 6 | 967 | 103 | ||
GenBank mRNA | AJ851638.1 | - | 5 - 6 | 929 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |