Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031310.1 | NP_001026481.1 | 8 - 9 | 852 | 76 | |
GenBank mRNA | AJ720946.1 | - | 8 - 9 | 852 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |