Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 13 - 14 | 1335 | 78 | |
XM_015293130.2 | XP_015148616.1 | 14 - 15 | 1580 | 78 | ||
XM_015293131.2 | XP_015148617.1 | 14 - 15 | 1452 | 78 | ||
XM_015293132.2 | XP_015148618.1 | 13 - 14 | 1513 | 78 | ||
GenBank mRNA | AJ868233.1 | - | 13 - 14 | 1335 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |