Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 5 - 6 | 631 | 104 | |
XM_015293130.2 | XP_015148616.1 | 6 - 7 | 876 | 104 | ||
XM_015293131.2 | XP_015148617.1 | 6 - 7 | 748 | 104 | ||
XM_015293132.2 | XP_015148618.1 | 6 - 7 | 878 | 104 | ||
GenBank mRNA | AJ868233.1 | - | 5 - 6 | 631 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |