Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033643.1 | NP_001028815.1 | 409 | 79 | ||
GenBank mRNA | AB109024.1 | - | 409 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus |