Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039255.1 | NP_001034344.1 | 2 - 3 | 280 | 81 | |
GenBank mRNA | AJ720244.1 | - | 2 - 3 | 280 | 81 | |
BX931383.1 | - | 275 | 81 | |||
BX932666.2 | - | 2 - 3 | 81 | |||
KU325499.1 | - | 1 - 2 | 99 | 81 | ||
KU557484.1 | - | 1 - 2 | 99 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |