Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039277.1 | NP_001034366.1 | 12 - 13 | 1743 | 65 | |
GenBank mRNA | AJ851760.1 | - | 12 - 13 | 1743 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |