Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039311.1 | NP_001034400.1 | 1 - 2 | 289 | 75 | |
GenBank mRNA | AJ719811.1 | - | 1 - 2 | 289 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |