Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039318.1 | NP_001034407.1 | 3 - 4 | 1153 | 79 | |
XM_015296936.2 | XP_015152422.1 | 8 - 9 | 3036 | 79 | ||
XM_015296939.2 | XP_015152425.1 | 7 - 8 | 2925 | 79 | ||
XM_025142546.1 | XP_024998314.1 | 8 - 9 | 3036 | 79 | ||
XM_025142547.1 | XP_024998315.1 | 7 - 8 | 2925 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |